missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Trypsin (Enzymatic Pretreatment) Buffer + 30mL Trypsin Concentrate

Product Code. 12623977 Shop All Epredia Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12623977 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12623977 Supplier Epredia™ Supplier No. TA125TR

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Epredia™ Lab Vision Trypsin (Enzymatic Pretreatment) is ideal for immunohistochemical procedures.

Trypsin is a digestive enzyme commonly used in formalin-fixed, paraffin-embedded immunohistochemical procedures. Its digestive properties are in many cases required as a pretreatment, prior to the primary antibody application, to produce a signal with certain protein-antibody interactions. It is found more useful than boiling pretreatments for antibodies such as AE3 and AE1/AE3.
  • Supplied as a two-component system

Includes:

TA-015-TR: 15mL (Buffer) (TA-015-TRB), 5mL (Trypsin concentrate) (TA-005-TRC)

TA-125-TR: 125mL (Buffer) (TA-125-TRB), 30mL (Trypsin concentrate) (TA-030-TRC)

Recommended for:

Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C.
Product Type Trypsin
Quantity 125 mL
Formulation 125mL Buffer + 30mL Trypsin concentrate
For Use With (Application) Immunohistochemistry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.