missing translation for 'onlineSavingsMsg'
Learn More

Tau Ab-2 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Tau Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia MS-247-P1ABX

Code : LD

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12680677

  • £370.00 / 100µL
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Tau proteins are microtubule-associated proteins (MAPs). Alternate splicing of tau mRNA and differential phosphorylation contribute to the heterogeneity of tau.

Host Species: Mouse

Clone: TAU-5

Isotype: IgG1

Species Reactivity: Human, Sheep, Cow, Mouse and Rat. Others not known.

Epitope: Middle of Tau

Immunogen: Purified bovine microtubule-associated proteins

Molecular Weight: 45-68kDa

Positive Control: IMR5 cells, Human T98G glioblastoma cells or Alzheimer's disease brain tissue

Cellular Localization: Cytoplasmic Ab-2 reacts with the non-phosphorylated as well as the phosphorylated forms of tau and its epitope maps in the middle of tau. Ab-2 intensely stains the human neurofibrillary tangles, neuropil threads, and neuritic plaques associated with Alzheimer's disease. Ab-2 shows no cross reaction with other MAPs or tubulin.

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

TAU Ab-2
Monoclonal
1mg/mL
Mouse
100 μL
Neuroscience
Bovine, Human, Mouse, Rat, Sheep
IgG1
Immunofluorescence, Immunoprecipitation, Western Blot
TAU-5
Unconjugated
Purified bovine microtubule-associated proteins
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers