missing translation for 'onlineSavingsMsg'
Learn More

Tau Ab-2 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Tau Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia MS-247-B1

Code : LD

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12640677

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Tau proteins are microtubule-associated proteins (MAPs). Alternate splicing of tau mRNA and differential phosphorylation contribute to the heterogeneity of tau.

Host Species: Mouse

Clone: TAU-5

Isotype: IgG1

Species Reactivity: Human, Sheep, Cow, Mouse and Rat. Others not known.

Epitope: Middle of Tau

Immunogen: Purified bovine microtubule-associated proteins

Molecular Weight: 45-68kDa

Positive Control: IMR5 cells, Human T98G glioblastoma cells or Alzheimer's disease brain tissue

Cellular Localization: Cytoplasmic Ab-2 reacts with the non-phosphorylated as well as the phosphorylated forms of tau and its epitope maps in the middle of tau. Ab-2 intensely stains the human neurofibrillary tangles, neuropil threads, and neuritic plaques associated with Alzheimer's disease. Ab-2 shows no cross reaction with other MAPs or tubulin.

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

TAU Ab-2
Monoclonal
200μg/mL
Mouse
500 μL
Neuroscience
Bovine, Human, Mouse, Rat, Sheep
IgG1
Immunofluorescence, Immunoprecipitation, Western Blot
TAU-5
Biotin
Purified bovine microtubule-associated proteins
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers