Learn More
Tau Ab-2 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Tau Ab-2, Mouse Monoclonal Antibody.
Brand: Epredia MS-247-B0
Code : LD
Additional Details : Weight : 0.00170kg
Description
Tau proteins are microtubule-associated proteins (MAPs). Alternate splicing of tau mRNA and differential phosphorylation contribute to the heterogeneity of tau.Host Species: Mouse
Clone: TAU-5
Isotype: IgG1
Species Reactivity: Human, Sheep, Cow, Mouse and Rat. Others not known.
Epitope: Middle of Tau
Immunogen: Purified bovine microtubule-associated proteins
Molecular Weight: 45-68kDa
Positive Control: IMR5 cells, Human T98G glioblastoma cells or Alzheimer's disease brain tissue
Cellular Localization: Cytoplasmic Ab-2 reacts with the non-phosphorylated as well as the phosphorylated forms of tau and its epitope maps in the middle of tau. Ab-2 intensely stains the human neurofibrillary tangles, neuropil threads, and neuritic plaques associated with Alzheimer's disease. Ab-2 shows no cross reaction with other MAPs or tubulin.
Recommended for:
- Immunohistochemistry (Not suitable)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
TAU Ab-2 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Neuroscience | |
Bovine, Human, Mouse, Rat, Sheep | |
IgG1 |
Immunofluorescence, Immunoprecipitation, Western Blot | |
TAU-5 | |
Biotin | |
Purified bovine microtubule-associated proteins | |
RUO | |
Primary | |
Concentrated |