missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Synaptophysin (Neuroendocrine Marker) Rabbit Monoclonal Antibody

Synaptophysin (Neuroendocrine Marker), Rabbit MAb is part of a comprehensive ab portfolio. Studies show rabbit mabs are more sensitive and specific than other available markers.

Brand:  Epredia™ RM-9111-S1

 View more versions of this product

Product Code. 12683717

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

This antibody recognizes a protein of 38kDa, identified as synaptophysin. It labels normal neuroendocrine cells of human adrenal medulla, carotid body, skin, pituitary gland, thyroid, lung, pancreas, gastrointestinal mucosa, Paneth's cells in the gastrointestinal tract and of gastric parietal cells. Neurons in the brain, spinal cord, and retina are also labeled. In combination with anti-chromogranin A and anti-NSE, Ab to synaptophysin is very useful in the identification of normal neuroendocrine cells and neuroendocrine neoplasms.

Host Species: Rabbit

Clone: SP11

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide of human synaptophysin

Molecular Weight: 38kDa

Positive Control: IMR5 cells, Pancreas or Pheochromocytoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Synaptophysin (Neuroendocrine Marker)
Monoclonal
Unconjugated
A synthetic peptide of human synaptophysin
IVD
Primary
Concentrated
Western Blot
SP11
Rabbit
500 μL
Neuroscience
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.