missing translation for 'onlineSavingsMsg'
Learn More

Synaptophysin Ab-4 Rabbit Polyclonal Antibody, Epredia™

Provide accurate, reproducible results Epredia™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-1461-P

Code : LD

Additional Details : Weight : 4.98950kg

 View more versions of this product

Product Code. 12642927

  • £422.00 / 1mL
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human synaptophysin

Molecular Weight: 38kDa

Positive Control: IMR-5 cells, Pancreas or Pheochromocytoma or GI tumor

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Synaptophysin Ab-4
Polyclonal
1mg/mL
Rabbit
1 mL
Cancer and Tumor Biology
Human
IgG
Western Blot
Polyclonal
Unconjugated
A synthetic peptide from C-terminus of human synaptophysin
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers