missing translation for 'onlineSavingsMsg'
Learn More

SV40 Large T Antigen Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ SV40 Large T Antigen Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-1832-PABX

 View more versions of this product

Product Code. 12630237

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

The simian virus 40 large T antigen plays important role in SV40 transfection and transformation of the cells. It binds to Rb and p53 suppressor proteins leading to the inactivation of suppressor pathway.

Host Species: Mouse

Clone: PAb101; originally clone 412

Isotype: IgG2a1

Species Reactivity: SV40 virus.

Epitope: C-terminal

Immunogen: SV40-transformed mouse line B4

Molecular Weight: 94kDa

Positive Control: SV40-transfected cells

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SV40 Large T Antigen Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Infectious Disease
Simian virus 40 (SV40)
Immunofluorescence, Immunoprecipitation, Western Blot
PAb101; same as clone 412
Unconjugated
SV40 transformed mouse line B4
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.