missing translation for 'onlineSavingsMsg'
Learn More

Smooth Muscle Actin Rabbit Polyclonal Antibody, Epredia™

Stain smooth muscle cells in vessel walls, gut wall, and myometrium with Epredia™ Actin, Smooth Muscle, Rabbit Polyclonal Antibody.

£106.00 - £377.00

Specifications

Antigen Actin, Smooth Muscle
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 4
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12623297
View Documents
Epredia
RB-9010-P0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12633297
View Documents
Epredia
RB-9010-P1
500 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12613297
View Documents
Epredia
RB-9010-P
1 mL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12653297
View Documents
Epredia
RB-9010-R7
7 mL Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This antibody stains smooth muscle cells in vessel walls, gut wall, and myometrium. Myoepithelial cells in breast and salivary gland are also stained. It reacts with tumors arising from smooth muscles and myoepithelial cells.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Baboon, Cow, Rabbit, Mouse, Rat and Chicken. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide from N-terminus of human α-smooth muscle isoform of actin

Positive Control: Smooth muscle of arteries, gut wall, uterus and Leiomyoma

Cellular Localization: Cytoplasmic.

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Actin, Smooth Muscle
Polyclonal
Rabbit
Muscle Biology
A synthetic peptide from N-terminus of human alpha-smooth muscle isoform of actin
Primary
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Bovine, Chicken, Human, Mouse, Non-human Primate, Rabbit
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.