missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ S100 Protein Ab-2 Rabbit Polyclonal Antibody with BSA and azide

Epredia™ S100 Protein Ab-2, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Brand:  Epredia™ RB-044-A

 View more versions of this product

Product Code. 12672737

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an α and β chain whereas S100B is composed of two β chains. Antibody to S100 stains Schwannomas, ependymomas, astrogliomas, almost all benign and malignant melanomas and their metastases. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Cow, Kangaroo, Mouse, Rat and Chicken. Others not tested.

Epitope: Not determined

Immunogen: S100 protein purified from cow brain

Molecular Weight: ∽22kDa (nonreduced); ∽11kDa (reduced)

Positive Control: Melanoma or Schwannoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunocytochemistry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

S100 Protein Ab-2
Polyclonal
Unconjugated
S100 protein purified from cow brain
1 mL
Cancer and Tumor Biology
Bovine, Chicken, Human, Kangaroo, Mouse
IgG
Immunohistochemistry (Paraffin)
Polyclonal
Rabbit
Melanoma or Schwannoma
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.