Learn More
Epredia™ Lab Vision™ RAD51 Ab-1 Mouse Monoclonal Antibody, biotin labeled with BSA and azide
Provide accurate, reproducible results with the Epredia™ RAD51 Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-988-P
Code : LD
Additional Details : Weight : 0.00100kg
Description
Rad51 (a eukaryotic homologue of Escherichia coli RecA) is similar to RecA, both biochemically and structurally: It promotes homologous pairing and strand exchange within a regular nucleoprotein filament. In yeast, the Rad51 family of related proteins also includes Rad55, Rad57 and Dmc1. In mammalian cells, seven genes in this family have been identified (HsRAD51, XRCC2, XRCC3, RAD51B/hREC2 and HsDMC1, RAD51C, and RAD 51D). Recent studies suggested that Rad51 is involved not only in homologous recombination but also in cell proliferation regulation.Host Species: Mouse
Clone: 51RAD01; same as 3C10
Isotype: IgG1/κ
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: Not determined
Immunogen: Recombinant RAD51 protein
Molecular Weight: 37-43kDa
Positive Control: HeLa cells, Testis
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
RAD51 Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
DNA Repair | |
Human, Mouse, Rat | |
IgG2a κ |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
51RAD01; same as 3C10 | |
Unconjugated | |
Recombinant Rad51 protein | |
RUO | |
Primary | |
Concentrated |