missing translation for 'onlineSavingsMsg'
Learn More

Prolactin Ab-2 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Prolactin Ab-2, Mouse Monoclonal Antibody.

£117.30 - £463.39

Specifications

Antigen Prolactin Ab-2
Clone PRL02
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
12619656
View Documents
Epredia
MS-1362-P0
100 μL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12649656
View Documents
Epredia
MS-1362-P1ABX
100 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12639656
View Documents
Epredia
MS-1362-PABX
200 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12629656
View Documents
Epredia
MS-1362-P1
500 μL Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
£342.00
£320.59 / 500µL
Save £21.41
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12609656
View Documents
Epredia
MS-1362-P
1 mL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12669656
View Documents
Epredia
MS-1362-R7
7 mL Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. Prolactin producing cells make up approximately 20% of the pituitary.

Host Species: Mouse

Clone: PRL02

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human prolactin

Molecular Weight: 23.5kDa

Positive Control: Pituitary gland

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • ELISA

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Prolactin Ab-2
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
IgG2a κ
PRL02
Monoclonal
Mouse
Steroids Hormones and Growth Factors
Human prolactin
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.