missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Plasma Cell Ab-1 Mouse Monoclonal Antibody, 50μg/mL, BSA and azide

Provide accurate, reproducible results with the Epredia™ Plasma Cell Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-488-P1

 View more versions of this product

Product Code. 12631567

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Host Species: Mouse

Clone: LIV3G11

Isotype: IgG2a

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Positive Control: Tonsil or lymph node

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Plasma Cell Ab-1
Monoclonal
50μg/mL
Mouse
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin)
LIV3G11
Unconjugated
500 μL
Hematopoietic
Human
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.