Learn More
Epredia™ Lab Vision™ Plasma Cell Ab-1 Mouse Monoclonal Antibody, 50μg/mL, BSA and azide
Provide accurate, reproducible results with the Epredia™ Plasma Cell Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-488-P
Description
Host Species: Mouse
Clone: LIV3G11
Isotype: IgG2a
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Positive Control: Tonsil or lymph node
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Note:
USA: RUO; Int'l: RUO
Specifications
| Plasma Cell Ab-1 | |
| Monoclonal | |
| 50μg/mL | |
| Mouse | |
| RUO | |
| Primary | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| LIV3G11 | |
| Unconjugated | |
| 1 mL | |
| Hematopoietic | |
| Human | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.