missing translation for 'onlineSavingsMsg'
Learn More

Placental Alk. Phosphatase (PLAP) Ab-1 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Placental Alk. Phosphatase (PLAP) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-208-P1

Code : LD

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12670547

  • £323.00 / 500µL
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Host Species: Mouse

Clone: 8B6.18

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP)

Molecular Weight: 60-70kDa

Positive Control: Placenta

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • ELISA

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Placental Alk. Phosphatase (PLAP) Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Cancer and Tumor Biology
Human
IgG2a κ
Immunohistochemistry (Paraffin)
8B6.18
Unconjugated
HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP)
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers