Learn More
Placental Alk. Phosphatase (PLAP) Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Placental Alk. Phosphatase (PLAP) Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-208-P1ABX
Description
Host Species: Mouse
Clone: 8B6.18
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP)
Molecular Weight: 60-70kDa
Positive Control: Placenta
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- ELISA
Note:
USA: RUO; Int'l: RUO
Specifications
| Placental Alk. Phosphatase (PLAP) Ab-1 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 100 μL | |
| Cancer and Tumor Biology | |
| Human | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| 8B6.18 | |
| Unconjugated | |
| HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP) | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.