Learn More
Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X)
Employ a wash buffer for immunolabeling techniques with the Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X).
Brand: Epredia™ TA-125-PT
Code : LD
Additional Details : Weight : 0.18000kg
Description
- For in vitro diagnostic use
- Added to promote more effective washings, resulting in decreased nonspecific background staining
- Aids reagent spreading on slides
Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week.
Specifications
PBS and Tween | |
Anatomical Pathology | |
Room Temperature |
125 mL | |
20X | |
7.6 |
For in vitro diagnostic use