missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X)

Employ a wash buffer for immunolabeling techniques with the Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X).

Brand:  Epredia™ TA-999-PT

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12364443

  • £184.00 / 1000mL
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • For in vitro diagnostic use
  • Added to promote more effective washings, resulting in decreased nonspecific background staining
  • Aids reagent spreading on slides

Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week.
Specifications

Specifications

PBS and Tween
Anatomical Pathology
Room Temperature
1000 mL
20X
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For in vitro diagnostic use