missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Pax-5 Rabbit Monoclonal Antibody

Take advantage of part of a comprehensive antibody portfolio with the Epredia™ Pax-5, Rabbit Monoclonal Antibody.

Brand:  Epredia™ RM-9133-R7

 View more versions of this product

Product Code. 12693797

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

PAX-5 is a B-cell-specific activator protein (BSAP). In the early stages of B cell developmemt, PAX-5 influences the expression of several B-cell-specific genes, such as CD19 and CD20. PAX-5 is expressed primarily in pro-, pre-, and mature B cells, but not in plasma cells. PAX-5 is expressed in most B-cell malignancies (pre-B and mature B cell lymphomas/leukemias). Over 90% of cases are positive for PAX-5 in Hodgkin's lymphoma, lymphoplasmacytic lymphoma, Merkel cell and small cell carcinoma. T-cell lymphomas, myeloma/plasmacytoma and carcinoid tumors are negative for PAX-5.

Host Species: Rabbit

Clone: SP34

Isotype: IgG

Species Reactivity: Human. Others not tested.

Immunogen: Synthetic peptide derived from the C-terminus of human PAX-5 protein

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PAX-5
Monoclonal
Unconjugated
Synthetic peptide derived from the C-terminus of human Pax-5 protein
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin)
SP34
Rabbit
7 mL
Cancer and Tumor Biology
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.