missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ PARP (Poly ADP-Ribose Polymerase) Ab-2 Rabbit Polyclonal Antibody with BSA and azide

Deliver accurate, reproducible results with the Epredia™ PARP (Poly ADP-Ribose Polymerase) Ab-2, Rabbit Polyclonal Antibody.

Brand:  Epredia™ RB-1516-P1ABX

 View more versions of this product

Product Code. 12682977

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide for the N-terminal region of human PARP

Molecular Weight: 116kDa

Positive Control: Raji cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PARP (Poly ADP-Ribose Polymerase) Ab-2
Polyclonal
Unconjugated
A synthetic peptide for the N-terminal region of human PARP
500 μL
Apoptosis
Human, Mouse
IgG
Western Blot
1mg/mL
Rabbit
Purified w/o BSA and Azide
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.