missing translation for 'onlineSavingsMsg'
Learn More

Pan Cytokeratin Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Cytokeratin, Pan Ab-1, Mouse Monoclonal Antibody.

£98.23 - £416.16

Specifications

Antigen Cytokeratin, Pan Ab-1
Clone AE1/AE3
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
12671087
View Documents
Epredia
MS-343-P0
100 μL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12651087
View Documents
Epredia
MS-343-P1ABX
100 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12681087
View Documents
Epredia
MS-343-PABX
200 μL Concentrated 1mg/mL
ONLINE EXCLUSIVE PRICE
£565.00
£355.53 / 200µL
Save £209.47
37% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12661087
View Documents
Epredia
MS-343-P1
500 μL Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
£199.00
£186.48 / 500µL
Save £12.52
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12641087
View Documents
Epredia
MS-343-P
1 mL Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
£412.00
£276.20 / 1mL
Save £135.80
32% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12601097
View Documents
Epredia
MS-343-R7
7 mL Prediluted Prediluted
ONLINE EXCLUSIVE PRICE
£172.00
£160.47 / 7mL
Save £11.53
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12611097
View Documents
Epredia
MS-343-RQ
12 mL Ready-to-Use Prediluted
ONLINE EXCLUSIVE PRICE
£257.00
£240.01 / 12mL
Save £16.99
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI <5.7) and basic (pI >6.0) subfamilies. The acidic keratins have molecular weights of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40 kDa. The basic keratins have molecular weights of 65-67, 64, 59, 58, 56 and 52 kDa. Members of acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with cell type, differentiation status and environment. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Host Species: Mouse

Clone: AE1/AE3

Isotype: IgG1 + IgG1

Species Reactivity: Human, Monkey, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.

Immunogen: Human epidermal keratin

Positive Control: Skin, Lung CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Specifications

Specifications

Cytokeratin, Pan Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human epidermal keratin
AE1/AE3
Monoclonal
Mouse
Cytoskeletal Protein
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.