missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Pan Cadherin Rabbit Polyclonal Antibody with BSA and azide

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cadherin, Pan, Rabbit Polyclonal Antibody.

Brand:  Epredia™ RB-9036-P0

 View more versions of this product

Product Code. 12603387

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Cadherins are calcium-dependent cell adhesion molecules, which play important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. In adhesion junctions, cadherins are bound to Beta and gamma catenins, which in turn bind to alpha catenin, an actin binding protein. Cadherins play important role in tumor invasion and metastasis. This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Chicken, Dog, Monkey, Cow and Xenopus.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of chicken N-cadherin

Molecular Weight: 120kDa

Positive Control: MCF-Cells, Tonsil and squamous epithelium

Cellular Localization: Membrane

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cadherin, Pan
Polyclonal
Unconjugated
A synthetic peptide from the C-terminus of chicken N-cadherin
RUO
Primary
Concentrated
Western Blot
200µg/mL
Rabbit
100 μL
Junction Protein
Bovine, Canine, Chicken, Human, Mouse, Non-human Primate, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.