missing translation for 'onlineSavingsMsg'
Learn More

p53 Ab-6 Mouse Monoclonal Antibody, Epredia™

Achieve accurate, reproducible results with the Epredia™ p53 Ab-6, Mouse Monoclonal Antibody.

Brand:  Epredia MS-187-P

Code : LD

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 12660377

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: DO-1

Isotype: IgG2a

Species Reactivity: Human, Monkey and Cow. Reacts weakly with Mouse and Rat. Others not known.

Epitope: aa 11-25

Immunogen: Recombinant human wild-type p53 protein expressed in E. coli

Molecular Weight: 53kDa

Positive Control: SKBR-3 cells or MDA-231 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry
  • Gel Supershift Assay

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

p53 Ab-6
Monoclonal
200μg/mL
Mouse
1 mL
Cell Cycle
Bovine, Human, Non-human Primate
IgG2a
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
DO-1
Unconjugated
Recombinant human wild-type p53 protein expressed in E. coli
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers