missing translation for 'onlineSavingsMsg'
Learn More

p53 Ab-5 Mouse Monoclonal Antibody, Epredia™

Achieve accurate, reproducible results with the Epredia™ p53 Ab-5, Mouse Monoclonal Antibody.

£98.43 - £510.00

Specifications

Antigen p53 Ab-5
Clone DO-7
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Mouse
View More Specs

Products 10
Product Code Brand Quantity Conjugate Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Conjugate Form Concentration Price Quantity & Availability  
12650357
missing translation for 'viewDocuments'
Epredia
MS-186-B0
100 μL Biotin Concentrated 200 μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12620357
missing translation for 'viewDocuments'
Epredia
MS-186-B1
500 μL Biotin Concentrated 200 μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12610357
missing translation for 'viewDocuments'
Epredia
MS-186-B
1 mL Biotin Concentrated 200 μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12630357
missing translation for 'viewDocuments'
Epredia
MS-186-P0
100 μL Unconjugated Concentrated 200 μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12690357
missing translation for 'viewDocuments'
Epredia
MS-186-P1ABX
100 μL Unconjugated Concentrated 1 mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12680357
missing translation for 'viewDocuments'
Epredia
MS-186-PABX
200 μL Unconjugated Concentrated 1 mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12660357
missing translation for 'viewDocuments'
Epredia
MS-186-P1
500 μL Unconjugated Concentrated 200 μg/mL
ONLINE EXCLUSIVE PRICE
£321.00
£301.45 / 500µL
Save £19.55
6% Off
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
12640357
missing translation for 'viewDocuments'
Epredia
MS-186-P
1 mL Unconjugated Concentrated 200 μg/mL
ONLINE EXCLUSIVE PRICE
£463.00
£436.11 / 1mL
Save £26.89
5% Off
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
12620367
missing translation for 'viewDocuments'
Epredia
MS-186-R7
7 mL Unconjugated Prediluted Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12630367
missing translation for 'viewDocuments'
Epredia
MS-186-RQ
12 mL Unconjugated Ready-to-Use Prediluted
ONLINE EXCLUSIVE PRICE
£257.00
£240.41 / 12mL
Save £16.59
6% Off
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: DO-7

Isotype: IgG2b/κ

Species Reactivity: Human, Monkey, Cow. Others not known.

Epitope: aa 37-45

Immunogen: Recombinant human wild-type p53 protein expressed in E. coli

Molecular Weight: 53kDa

Positive Control: SKBR-3 or MDA-231 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • Flow Cytometry

Specifications

Specifications

p53 Ab-5
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Mouse
Cancer and Tumor Biology
Primary
DO-7
Monoclonal
IVD
Recombinant human wild-type p53 protein expressed in E. coli
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.