missing translation for 'onlineSavingsMsg'
Learn More

p21 WAF1 Ab-11 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results with the Epredia™ p21WAF1 Ab-11, Mouse Monoclonal Antibody.

Brand:  Epredia MS-891-B1

Code : LD

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12632547

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

p21WAF1/Cip1/Sdi1/Pic1 is a tumor suppressor protein. Expression of p21WAF1 is induced by wild type, but not mutant, p53 suppressor protein. The p21WAF1 protein binds to cyclin/CDK complexes and inhibits their kinase activity thereby stopping cell cycle progression. It also binds to PCNA (proliferating cell nuclear antigen) and blocks DNA replication but not the DNA repair process.

Host Species: Mouse

Clone: CP74

Isotype: IgG2b

Species Reactivity: Human and Rat. Others not known.

Epitope: aa 1-80

Immunogen: Full-length human recombinant p21 protein

Molecular Weight: 21kDa

Positive Control: Raji, PC12 cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

p21superscriptWAF1superscript Ab-11
Monoclonal
200μg/mL
Mouse
500 μL
Cell Cycle
Human, Rat
IgG2b
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
CP74
Biotin
Full length human recombinant p21 protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers