Learn More
p18INK4c Ab-3 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results with the Epredia™ p18INK4c Ab-3, Mouse Monoclonal Antibody.
Brand: Epredia MS-858-PABX
Description
p18INK4c is a specific inhibitor of cdk4/cdk6 and is expressed during G1 -to S transition in the eukaryotic cell division cycle. It is maximally induced as cells enter S-phase.Host Species: Mouse
Clone: 18P118; same as DCS-118
Isotype: IgG1
Species Reactivity: Human, Mouse. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant p18INK4c protein
Molecular Weight: 18kDa
Positive Control: HeLa cells; Tonsil, skeletal muscle, heart, pancreas
Cellular Localization: Cytoplasmic and Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
| p18superscriptINK4csuperscript Ab-3 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 200 μL | |
| Cell Cycle | |
| Human, Mouse | |
| IgG1 |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 18P118; same as DCS-118 | |
| Unconjugated | |
| Purified recombinant p18superscriptINK4csuperscript protein | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.