missing translation for 'onlineSavingsMsg'
Learn More

p18INK4c Ab-3 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results with the Epredia™ p18INK4c Ab-3, Mouse Monoclonal Antibody.

Brand:  Epredia MS-858-P0

 View more versions of this product

Product Code. 12662457

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

p18INK4c is a specific inhibitor of cdk4/cdk6 and is expressed during G1 -to S transition in the eukaryotic cell division cycle. It is maximally induced as cells enter S-phase.

Host Species: Mouse

Clone: 18P118; same as DCS-118

Isotype: IgG1

Species Reactivity: Human, Mouse. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant p18INK4c protein

Molecular Weight: 18kDa

Positive Control: HeLa cells; Tonsil, skeletal muscle, heart, pancreas

Cellular Localization: Cytoplasmic and Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

p18superscriptINK4csuperscript Ab-3
Monoclonal
200μg/mL
Mouse
100 μL
Cell Cycle
Human, Mouse
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
18P118; same as DCS-118
Unconjugated
Purified recombinant p18superscriptINK4csuperscript protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.