missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Nucleophosmin (NPM) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ Nucleophosmin (NPM) Ab-1, Mouse Monoclonal Antibody.

£106.08 - £473.28

Specifications

Antigen Nucleophosmin (NPM) Ab-1
Clone NA24
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
12660297
View Documents
Epredia™
MS-1849-P0
100 μL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12680297
View Documents
Epredia™
MS-1849-P1ABX
100 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12690297
View Documents
Epredia™
MS-1849-PABX
200 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12670297
View Documents
Epredia™
MS-1849-P1
500 μL Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
£357.00
£335.99 / 500µL
Save £21.01
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12650297
View Documents
Epredia™
MS-1849-P
1 mL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12620307
View Documents
Epredia™
MS-1849-R7
7 mL Prediluted Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Nucleophosmin (NPM)/B23/numatrin is a phosphoprotein associated with cell nucleoli. Treatment of cells with cytotoxic drugs may make nucleophosmin enter nucleoplasm. It is reported that nucleophosmin shuttles continuously between cytoplasm and nucleoli to carry newly synthesized ribosomal proteins into nucleoli. NPM-ALK is a hybrid of the anaplastic lymphoma kinase (ALK) gene and the nucleophosmin (NPM) gene resulting from the t(2;5)(p23;q35) translocation found in anaplastic large cell lymphoma. In acute promyelocytic leukemia NPM gene is fused to RARalpha gene in (5;17) (q33;q12) transition.

Host Species: Mouse

Clone: NA24

Isotype: IgG/κ

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: GST fusion protein containing the N-terminal part of nucleophosmin fused to 14aa of ALK protein

Molecular Weight: 38kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Nucleophosmin (NPM) Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
IgG κ
NA24
Monoclonal
Mouse
Cancer and Tumor Biology
GST fusion protein containing the N-terminal part of nucleophosmin fused to 14 aa of ALK protein
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.