missing translation for 'onlineSavingsMsg'
Learn More

Negative Control for Rabbit IgG Ab-1, Epredia™

Take advantage of a quality product that delivers accurate results with the Epredia™ Negative Control for Rabbit IgG Ab-1, Rabbit Polyclonal Antibody.

Brand:  Epredia NC-100-P1

 View more versions of this product

Product Code. 12692657

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

This IgG was purified from serum collected from a rabbit prior to immunization. It does not react with any antigen of human or animal cells. The serum was purified under the same experimental conditions as for the other rabbit polyclonal antibodies of NeoMarkers. In order to serve as a true negative control, this IgG should be used under the identical conditions and at the same concentration as NeoMarkers' other purified rabbit polyclonal IgG antibodies.

Host Species: Rabbit

Isotype: IgG

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Negative Control for Rabbit IgG Ab-1
Polyclonal
1mg/mL
Purified with BSA and Azide
500 μL
Negative Controls
All species
IgG
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Polyclonal
Unconjugated
Rabbit
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.