Learn More
Negative Control for Rabbit IgG Ab-1, Epredia™
Take advantage of a quality product that delivers accurate results with the Epredia™ Negative Control for Rabbit IgG Ab-1, Rabbit Polyclonal Antibody.
Brand: Epredia NC-100-R7
Description
This IgG was purified from serum collected from a rabbit prior to immunization. It does not react with any antigen of human or animal cells. The serum was purified under the same experimental conditions as for the other rabbit polyclonal antibodies of NeoMarkers. In order to serve as a true negative control, this IgG should be used under the identical conditions and at the same concentration as NeoMarkers' other purified rabbit polyclonal IgG antibodies.Host Species: Rabbit
Isotype: IgG
Recommended for:
Negative control for:
- Immunohistochemistry (Frozen and formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
- Flow Cytometry
- ELISA
- Gel Supershift
Note:
USA: RUO; Int'l: RUO
Specifications
| Negative Control for Rabbit IgG Ab-1 | |
| Polyclonal | |
| — | |
| Ready-to-use | |
| 7 mL | |
| Negative Controls | |
| All species | |
| IgG |
| ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| Primary | |
| Prediluted |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.