Learn More
Epredia™ MultiVision Polymer Detection System
Shop All Epredia ProductsDescription
- A primary antibody cocktail of choice may be used in conjunction with ready-to-use MultiVision polymer cocktail, composed of anti-Mouse/alkaline phosphatase (AP) and anti-Rabbit/horseradish peroxidase (HRP)
- Cocktail is specially formulated from MultiVision polymers that provide increased sensitivity, time-savings, and detection simplicity
- Yields higher sensitivity and antibody efficiency as well as better signal-to-noise ratios
- Biotin-free, eliminating background staining
Antigen Specificity: anti-Mouse IgG (H+L) and anti-Rabbit IgG (H+L)
Enzyme: Peroxidase and Phosphatase
Chromogen/Substrate: LVBlue/ LVRed
Includes:
TL-012-MARH: 1 MultiVision anti-rabbit/HRP + anti-mouse/AP polymer (cocktail) (TL-012-MAM), 1 Ultra V block (TA-012-UB), 1 Hydrogen Peroxidase Block (TA-012-HP), 1 LVBlue AP Buffer (TA-075-TMM), 1 LVBlue Chromogen Vial 1 (TA-001-AB1), 1 LVBlue Chromogen Vial 2 (TA-001-AB2), 1 LVBlue Chromogen Vial 3 (TA-001-AB3), 1 LVRed Chromogen Vial 1 (TA-004-HR1), 1 LVRed Chromogen Vial 2 (TA-002-HR2), 1 LVRed Chromogen Vial 3 (TA-002-HR3), 1 LVRed Chromogen Vial 4 (TA-002-HR4), 1 Mounting Media (TA-005-UA)
Note:
MultiVision Polymer Detection System may be employed with any primary antibody cocktail to visualize the presence of two proteins simultaneously. After the specific antibodies are bound to their respective antigens in tissue sections, the rabbit and mouse primary antibodies are located by a specially formulated cocktail of secondary antibody polymers: MultiVision anti-rabbit/HRP + MultiVision anti-mouse/AP. In general, the amino acid polymer is conjugated to AP or HRP enzymes and the Fab fragments of goat anti-rabbit or goat anti-mouse immunoglobulins. The polymer complex is then visualized with first the LVBlue chromogen for AP activity and after washing with the LVRed chromogen for HRP activity. A weak hematoxilin nuclear counterstain is not recommended.
Specifications
Specifications
| Chromagen | LVBlue |
| Enzyme | Peroxidase, Phosphatase |
| Product Line | Lab Vision™ |
| Specificity | anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) |
| Substrate | LVRed |
| Includes | TL-012-MARH: 1 MultiVision anti-rabbit/HRP + anti-mouse/AP polymer (cocktail) (TL-012-MAM), 1 Ultra V block (TA-012-UB), 1 Hydrogen Peroxidase Block (TA-012-HP), 1 LVBlue AP Buffer (TA-075-TMM), 1 LVBlue Chromogen Vial 1 (TA-001-AB1), 1 LVBlue Chromogen Vial 2 (TA-001-AB2), 1 LVBlue Chromogen Vial 3 (TA-001-AB3), 1 LVRed Chromogen Vial 1 (TA-004-HR1), 1 LVRed Chromogen Vial 2 (TA-002-HR2), 1 LVRed Chromogen Vial 3 (TA-002-HR3), 1 LVRed Chromogen Vial 4 (TA-002-HR4), 1 Mounting Media (TA-005-UA) |
| Quantity | 12 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.