Learn More
MMP-2 (72kDa Collagenase IV) Ab-2 Mouse Monoclonal Antibody, Epredia™
Description
Host Species: Mouse
Clone: VB3
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Human native 72kDa MMP-2
Molecular Weight: 72kDa (proform) and ∽66kDa (active form)
Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Not Suitable)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | MMP-2 (72kDa Collagenase IV) Ab-2 |
| Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | VB3 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | Human native 72kDa MMP-2 |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.