Learn More
Epredia™ Lab Vision™ MMP-11 (Stromelysin-3) Ab-5 Mouse Monoclonal Antibody, 200μg/mL, biotin labeled with BSA and azide
Ensure accurate, reproducible results in experiments with the Epredia™ MMP-11 (Stromelysin-3) Ab-5, Mouse Monoclonal Antibody.
£132.60 - £606.90
Specifications
| Antigen | MMP-11 (Stromelysin-3) Ab-5 |
|---|---|
| Clone | SL3.05 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Conjugate | Form | Concentration | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Conjugate | Form | Concentration | Price | Quantity & Availability | ||
|
12668676
|
Epredia™
MS-1035-B0 |
100 μL | Biotin | Concentrated | 200μg/mL | ||||
|
12698676
|
Epredia™
MS-1035-P0 |
100 μL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12628686
|
Epredia™
MS-1035-P1ABX |
100 μL | Unconjugated | Concentrated | 1mg/mL | ||||
|
12618686
|
Epredia™
MS-1035-PABX |
200 μL | Unconjugated | Concentrated | 1mg/mL | ||||
|
12608686
|
Epredia™
MS-1035-P1 |
500 μL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12688676
|
Epredia™
MS-1035-P |
1 mL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12658686
|
Epredia™
MS-1035-R7 |
7 mL | Unconjugated | Prediluted | |||||
Description
Stromelysin, a member of the matrix metalloproteinase family, demonstrates wide substrate specificity with the ability to degrade proteoglycan, fibronectin, laminin, casein, and the nonhelical region of collagen. The stromelysin-3 (MMP-11) gene was originally identified on the basis that it is expressed specifically in stromal cells surrounding invasive breast carcinomas. MMP-11 is localized by in situ hybridization to the long arm of chromosome 22. The MMP-11 expression is found more specifically in malignant tumors than in benign ones.Host Species: Mouse
Clone: SL3.05
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Immunogen: Recombinant protein corresponding to active form of human MMP-11
Molecular Weight: ∽60kDa (latent), ∽47kDa (active) and ∽35kDa (degradation product)
Positive Control: Recombinant MMP-11 protein; Placenta
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native verified)
- Affinity Purification (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
| MMP-11 (Stromelysin-3) Ab-5 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Mouse | |
| Proteases And Inhibitors | |
| Recombinant protein corresponding to active form of human MMP-11 | |
| Primary |
| SL3.05 | |
| Monoclonal | |
| RUO | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.