Learn More
Mitochondria Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ Mitochondria Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia MS-1372-P
Code : LD
Additional Details : Weight : 0.00100kg
Description
Host Species: Mouse
Clone: MTC02
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Semi-purified mitochondrial preparation
Molecular Weight: 60kDa
Positive Control: HeLa cells, Liver
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Mitochondria Ab-2 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Cell Cycle | |
Human | |
IgG1 |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
MTC02 | |
Unconjugated | |
Semi-purified mitochondrial preparation | |
RUO | |
Primary | |
Concentrated |