Learn More
Epredia™ Lab Vision™ Mesothelioma Ab-1 Mouse Monoclonal Antibody
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Mesothelioma Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1494-R7
Code : LD
Additional Details : Weight : 0.01200kg
Description
Host Species: Mouse
Clone: HBME-1
Isotype: IgM/κ
Species Reactivity: Human. Others not known.
Immunogen: A suspension of human mesotheliema cells from patients with malignant epithelial mesothelioma
Positive Control: Mesothelioma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Mesothelioma Ab-1 | |
Monoclonal | |
Unconjugated | |
A suspension of human mesotheliema cells from patients with malignant epithelial mesothelioma | |
IVD | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
HBME-1 | |
Mouse | |
7 mL | |
Cancer and Tumor Biology | |
Human | |
IgM κ |