missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Mesothelin Ab-1 Mouse Monoclonal Antibody

Product Code. 12689556 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Unit Size:
100µL
1mL
500µL
7mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form unitSize
12689556 500 μL Concentrated 500µL
12679556 100 μL Concentrated 100µL
12649556 1 mL Concentrated 1mL
12659556 7 mL Prediluted 7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12689556 Supplier Epredia™ Supplier No. MS1320S1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Mesothelin Ab-1, Mouse Monoclonal Antibody.

Mesothelin is a glycocylphosphotidylianositol-linked cell-surface glycoprotein, that is present on the surface of normal mesothelium and is overexpressed in many patients with epithelial ovarian cancer and malignant mesotheliomas.

Host Species: Mouse

Clone: 5B2

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant protein corresponding to the membrane-bound form of the mesothelin molecule

Molecular Weight: 40kDa

Positive Control: Mesothelioma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mesothelin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5B2
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to the membrane-bound form of the mesothelin molecule
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.