missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Mayer's Hematoxylin

Product Code. 12603967 Shop All Epredia Products
Click to view available options
Quantity:
60 mL
125mL
Unit Size:
125mL
60mL
This item is not returnable. View return policy

Product Code. 12603967

Brand: Epredia™ TA125MH

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for use in histological applications, Epredia™ Lab Vision™ Mayer's Hematoxylin provides superior quality.

  • Requires slightly longer application periods
  • Based on the original formulation developed by Mayer
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature
Product Type Hematoxylin
Quantity 125mL
compliance-icons
Product Identifier
  • Mayer's Hematoxylin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H330-Fatal if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P320-Specific treatment is urgent (see on this label).
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : glycerin, acetic acid

For in vitro diagnostic use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.