missing translation for 'onlineSavingsMsg'
Learn More

MART-1/Melan-A (Melanoma Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ MART-1/Melan-A (Melanoma Marker) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-612-R7

 View more versions of this product

Product Code. 12661777

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.

Host Species: Mouse

Clone: M2-7C10

Isotype: IgG2b/κ

Species Reactivity: Human and Horse. Does not react with mouse and rat. Others not tested.

Epitope: Not determined

Immunogen: Recombinant hMART-1 protein

Molecular Weight: 18kDa

Positive Control: CAC1 melanoma cells and melanomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MART-1/Melan-A (Melanoma Marker) Ab-1
Monoclonal
Mouse
7 mL
Cancer and Tumor Biology
Equine, Human
IgG2b κ
Immunoprecipitation
M2-7C10
Unconjugated
Recombinant hMART-1 protein
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.