missing translation for 'onlineSavingsMsg'
Learn More

LCK (p56lck) Ab-1 Mouse Monoclonal Antibody, Epredia™

Recommended for Western Blotting, Immunoprecipitation (Denatured verified) and Kinase Assays, Epredia™ LCK (p56lck) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

£119.34 - £525.30

Specifications

Antigen LCK (p56superscriptlcksuperscript) Ab-1
Clone 3A5
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Product Code Brand Quantity Concentration Price Quantity & Availability  
Product Code Brand Quantity Concentration Price Quantity & Availability  
12600037
View Documents
Epredia
MS-164-P0
100 μL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12610037
View Documents
Epredia
MS-164-P1
500 μL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12620037
View Documents
Epredia
MS-164-P1ABX
100 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12630037
View Documents
Epredia
MS-164-PABX
200 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

p56lck is a membrane-associated non-receptor protein tyrosine kinase from the Src-family of kinases. The phosphorylation status and the activity of p56lck is regulated by the CD45 tyrosine protein phosphatase. p56lck plays a critical role in T cell development and activation. p56lck gene is localized to a site in the genome which undergoes frequent chromosomal abnormalities in lymphomas and neuroblastomas.

Host Species: Mouse

Clone: 3A5

Isotype: IgG2b/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 1-225

Immunogen: Recombinant protein corresponding to aa 1-225 of murine p56lck protein

Molecular Weight: 56kDa

Positive Control: Jurkat cells

Cellular Localization: Cell membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

LCK (p56superscriptlcksuperscript) Ab-1
Immunoprecipitation, Western Blot
Unconjugated
Mouse
Cancer and Tumor Biology
Recombinant protein corresponding to aa 1-225 of murine p56superscriptlcksuperscript protein
Primary
3A5
Monoclonal
Concentrated
RUO
Human, Mouse, Rat
IgG2b κ
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.