Learn More
Laminin B2/γ1 Ab-1 Rat Monoclonal Antibody, Epredia™
Recommended for Immunohistochemistry (Acetone-fixed frozen and acid-alcohol-fixed, paraffin-embedded), Immunofluorescence and Immunoprecipitation (Native and denatured), Epredia™ Laminin B2/γ1 Ab-1, Rat Monoclonal Antibody provides accurate, reproducible results.
£119.34 - £606.00
Specifications
| Antigen | Laminin B2/gamma1 Ab-1 |
|---|---|
| Clone | A5 |
| Applications | Immunohistochemistry (Frozen), Immunoprecipitation |
| Classification | Monoclonal |
| Host Species | Rat |
| Product Code | Brand | Quantity | Conjugate | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Conjugate | Concentration | Price | Quantity & Availability | |||
|
12613887
|
Epredia
RT-795-B0 |
100 μL | Biotin | 200µg/mL | |||||
|
12623887
|
Epredia
RT-795-B1 |
500 μL | Biotin | 200µg/mL | |||||
|
12633887
|
Epredia
RT-795-P0 |
100 μL | Unconjugated | 200µg/mL | |||||
|
12653887
|
Epredia
RT-795-P1 |
500 μL | Unconjugated | 200µg/mL | |||||
|
12643887
|
Epredia
RT-795-P |
1 mL | Unconjugated | 200µg/mL | |||||
|
12663887
|
Epredia
RT-795-P1ABX |
100 μL | Unconjugated | 1mg/mL | |||||
|
12673887
|
Epredia
RT-795-PABX |
200 μL | Unconjugated | 1mg/mL | |||||
Description
Laminins are large heterotrimeric, noncollagenous glycoproteins composed of α, β, and γ chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.Host Species: Rat
Clone: A5
Isotype: IgG2a/κ
Species Reactivity: Human and Mouse. Others not known.
Epitope: Not determined
Immunogen: Murine EHS laminin preparation
Molecular Weight: ∽210kDa
Positive Control: LS174T cells, Normal colon or colon CA
Cellular Localization: Basement membrane Specificity of Ab-1 was established by immunoprecipitation and immunofluorescence of human skeletal muscle and kidney with laminin chain-specific MAbs. Ab-1 stains uniformly all human and murine basement membranes.
Recommended for:
- Immunohistochemistry (Acetone-fixed frozen and acid-alcohol-fixed, paraffin-embedded)
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
| Laminin B2/gamma1 Ab-1 | |
| Immunohistochemistry (Frozen), Immunoprecipitation | |
| Rat | |
| Extracellular Matrix | |
| Murine EHS laminin preparation | |
| Primary |
| A5 | |
| Monoclonal | |
| RUO | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.