Learn More
L1 Cell Adhesion Molecule Ab-1 Mouse Monoclonal Antibody, Epredia™
Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation (Native and denatured), Epredia™ L1 Cell Adhesion Molecule Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
£116.00 - £655.00
Specifications
| Antigen | L1 Cell Adhesion Molecule Ab-1 |
|---|---|
| Clone | UJ127 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Conjugate | Form | Concentration | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Conjugate | Form | Concentration | Price | Quantity & Availability | ||
|
12652287
|
Epredia
MS-770-B0 |
100 μL | Biotin | Concentrated | 200μg/mL | ||||
|
12662287
|
Epredia
MS-770-B1 |
500 μL | Biotin | Concentrated | 200μg/mL | ||||
|
12642287
|
Epredia
MS-770-B |
1 mL | Biotin | Concentrated | 200μg/mL | ||||
|
12682287
|
Epredia
MS-770-P0 |
100 μL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12612297
|
Epredia
MS-770-P1ABX |
100 μL | Unconjugated | Concentrated | 1mg/mL | ||||
|
12602297
|
Epredia
MS-770-PABX |
200 μL | Unconjugated | Concentrated | 1mg/mL | ||||
|
12692287
|
Epredia
MS-770-P1 |
500 μL | Unconjugated | Concentrated | 200μg/mL |
£390.00 £366.79 / 500µL Save £23.21 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
|
12672287
|
Epredia
MS-770-P |
1 mL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12662297
|
Epredia
MS-770-R7 |
7 mL | Unconjugated | Prediluted | Prediluted | ||||
Description
The L1CAM gene, which is located in Xq28, is involved in three distinct conditions: 1) HSAS (hydrocephalus-stenosis of the aqueduct of Sylvius); 2) MASA (mental retardation, aphasia, shuffling gait, adductus thumbs); and 3) SPG1 (spastic paraplegia). The L1, neural cell adhesion molecule (L1CAM) also plays an important role in axon growth, fasciculation, neural migration and in mediating neuronal differentiation. Expression of L1 protein is restricted to tissues arising from neuroectoderm.Host Species: Mouse
Clone: UJ127
Isotype: IgG1/k
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Homogenous suspension of 16-week human fetal brain
Molecular Weight: 220-240kDa
Positive Control: IMR-5 cells Neuroblastoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
| L1 Cell Adhesion Molecule Ab-1 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Mouse | |
| Cancer and Tumor Biology | |
| Homogenous suspension of 16 week human fetal brain | |
| Primary |
| UJ127 | |
| Monoclonal | |
| RUO | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.