Learn More
Ki-67 Ab-4 Rabbit Polyclonal Antibody, Epredia™
Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ Ki-67 Ab-4, Rabbit Polyclonal Antibody provides accurate, reproducible results. Rabbit Polyclonal Antibody.
Brand: Epredia RB-1510-RQ
Description
Ki67 is a nuclear protein, which is expressed in the proliferating cells. Ki67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiscent) phase are negative for this protein.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Rat. Others not known.
Epitope: Middle of Ki-67 protein
Immunogen: A synthetic peptide from the human Ki-67 protein
Positive Control: Tonsil or Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| Ki-67 Ab-4 | |
| Polyclonal | |
| Unconjugated | |
| A synthetic peptide from the human Ki67 protein | |
| RUO | |
| Primary |
| Immunohistochemistry (Paraffin) | |
| Prediluted | |
| Rabbit | |
| 12 mL | |
| Cancer and Tumor Biology | |
| Ready-to-Use |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.