missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Involucrin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Epredia™ Involucrin Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

£104.04 - £413.10

Specifications

Antigen Involucrin Ab-1
Clone SY5
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Mouse
View More Specs

Products 6
Product Code Brand Quantity Conjugate Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Conjugate Form Concentration Price Quantity & Availability  
12649416
View Documents
Epredia™
MS-126-P0
100 μL Unconjugated Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12679416
View Documents
Epredia™
MS-126-P1ABX
100 μL Unconjugated Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12689416
View Documents
Epredia™
MS-126-PABX
200 μL Unconjugated Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12669416
View Documents
Epredia™
MS-126-P1
500 μL Unconjugated Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
£335.00
£314.63 / 500µL
Save £20.37
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12659416
View Documents
Epredia™
MS-126-P
1 mL Unconjugated Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12629426
View Documents
Epredia™
MS-126-R7
7 mL Unconjugated Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Involucrin is expressed in a range of stratified squamous epithelia, including the cornea. In normal epidermis, it is first expressed in the upper spinous layers, and in keratinocyte cultures it is expressed by all cells that have left the basal layer. Involucrin expression is abnormal in squamous cell carcinomas and premalignant lesions, and is reduced in severe dysplasias of the larynx and cervix.

Host Species: Mouse

Clone: SY5

Isotype: IgG1

Species Reactivity: Human, Gorilla, Owl, Monkey, Pig, Dog and Rat. Others not known.

Epitope: Codon 421-568

Immunogen: Human keratinocytes' involucrin

Molecular Weight: 66-170kDa

Positive Control: LS174T cells, Skin

Cellular Localization: Cytoplasmic Ab-1 stains the involucrin in a variety of sizes: 170kDa in MCF-7 cells, a doublet of ∽115kDa and 150kDa in gorilla and owl monkey, 66kDa in dog, and a doublet of 105kDa in pig.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • ELISA

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Involucrin Ab-1
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Mouse
Cytoskeletal Protein
Human keratinocytes involucrin
Primary
SY5
Monoclonal
RUO
Canine, Human, Non-human Primate, Pig, Rat
IgG1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.