missing translation for 'onlineSavingsMsg'
Learn More

IGF-1R (α-subunit) Ab-4 Mouse Monoclonal Antibody, Epredia™

Epredia™ IGF-1R (α-Subunit) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-644-P

 View more versions of this product

Product Code. 12621897

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of α- and Β-subunits, which are disulfide-linked in a Β-α-α-Β configuration in the mature receptor. The α-subunit is completely extracellular, while the Β-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.

Host Species: Mouse

Clone: 24-60

Isotype: IgG2a/κ

Species Reactivity: Human. Weakly reacts with Rabbit and Pig (Ab-2 is better). Does not react with rat.

Epitope: aa 184-283

Immunogen: IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA

Molecular Weight: 125kDa

Positive Control: Placenta or Breast CA

Cellular Localization: Membrane Ab-4 is excellent for immunoprecipitation of alpha-subunit of IGF-1R.

Recommended for:

  • Western Blotting (Not suitable)
  • Immunoprecipitation
  • Acts as a Weak IGF-like Agonist in Bioassays (Thymidine incorporation into DNA)
  • Substantially (>80%) Inhibits IGF-1 Binding

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IGF-1R (alpha-Subunit) Ab-4
Monoclonal
200μg/mL
Mouse
1 mL
Neuroscience
Human
IgG2a κ
Immunoprecipitation
24-60
Unconjugated
IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.