Learn More
IGF-1R (α-subunit) Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ IGF-1R (α-Subunit) Ab-1, Mouse Monoclonal Antibody.
£119.34 - £688.50
Specifications
| Antigen | IGF-1R (alpha-Subunit) Ab-1 |
|---|---|
| Clone | 24-31 |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Form | Concentrated |
| Product Code | Brand | Quantity | Conjugate | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Conjugate | Concentration | Price | Quantity & Availability | |||
|
12651877
|
Epredia
MS-641-B0 |
100 μL | Biotin | 200μg/mL | |||||
|
12661877
|
Epredia
MS-641-B1 |
500 μL | Biotin | 200μg/mL | |||||
|
12641877
|
Epredia
MS-641-B |
1 mL | Biotin | 200μg/mL | |||||
|
12681877
|
Epredia
MS-641-P0 |
100 μL | Unconjugated | 200μg/mL | |||||
|
12691877
|
Epredia
MS-641-P1 |
500 μL | Unconjugated | 200μg/mL |
£385.00 £335.05 / 500µL Save £49.95 12% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
|
12671877
|
Epredia
MS-641-P |
1 mL | Unconjugated | 200μg/mL | |||||
|
12601887
|
Epredia
MS-641-P1ABX |
100 μL | Unconjugated | 1mg/mL | |||||
|
12611887
|
Epredia
MS-641-PABX |
200 μL | Unconjugated | 1mg/mL | |||||
Description
IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of α- and Β-subunits, which are disulfide-linked in a Β-α-α-Β configuration in the mature receptor. The α-subunit is completely extracellular, while the Β-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.Host Species: Mouse
Clone: 24-31
Isotype: IgG1
Species Reactivity: Human. Weakly reacts with Rabbit. Does not react with rat. Pig unknown.
Epitope: aa 283-440 (exon 4-6)
Immunogen: IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA
Molecular Weight: 125kDa
Positive Control: Placenta or Breast CA
Cellular Localization: Membrane
Recommended for:
- Western Blotting (Not suitable)
- Immunoprecipitation
- ELISA
- Acts as a Weak IGF-like Agonist in Bioassays (Thymidine incorporation into DNA)
- Does Not Significantly Inhibit IGF-1 Binding
Note:
USA: RUO; Int'l: RUO
Specifications
| IGF-1R (alpha-Subunit) Ab-1 | |
| Immunoprecipitation | |
| Concentrated | |
| RUO | |
| Human | |
| IgG1 |
| 24-31 | |
| Monoclonal | |
| Mouse | |
| Neuroscience | |
| IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.