Learn More
Epredia™ Lab Vision™ IgD (δ-Heavy Chain) Ab-1 Rabbit Polyclonal Antibody
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ IgD (δ-Heavy Chain) Ab-1, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Brand: Epredia™ RB-1436-A0
Code : LD
Additional Details : Weight : 0.00170kg
Description
The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: IgD isolated from a pool of normal human sera
Molecular Weight: 50kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
IgD (delta-Heavy Chain) Ab-1 | |
Polyclonal | |
Unconjugated | |
IgD isolated from a pool of normal human sera | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
Polyclonal | |
Rabbit | |
100 μL | |
Immunoglobulins | |
Human | |
IgG |