missing translation for 'onlineSavingsMsg'
Learn More

Helicobacter pylori Rabbit Polyclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Helicobacter pylori, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-9070-R7

 View more versions of this product

Product Code. 12673447

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Helicobacter pylori is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Helicobacter pylori

Epitope: Not determined

Immunogen: Purified Helicobacter pylori

Positive Control: Helicobacter pylori infected tissue

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Helicobacter pylori
Polyclonal
Unconjugated
Purified Helicobacter pylori
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin)
Polyclonal
Rabbit
7 mL
Infectious Disease
H. pylori
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.