Learn More
Helicobacter pylori Rabbit Polyclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Helicobacter pylori, Rabbit Polyclonal Antibody.
Brand: Epredia RB-9070-P
Description
Helicobacter pylori is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Helicobacter pylori
Epitope: Not determined
Immunogen: Purified Helicobacter pylori
Positive Control: Helicobacter pylori infected tissue
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Helicobacter pylori | |
| Polyclonal | |
| 200µg/mL | |
| Rabbit | |
| 1 mL | |
| Infectious Disease | |
| H. pylori | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Polyclonal | |
| Unconjugated | |
| Purified Helicobacter pylori | |
| IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.