Learn More
Epredia™ Lab Vision™ Human Papilloma Virus (HPV) Ab-3 Mouse Monoclonal Antibody
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Human Papilloma Virus (HPV) Ab-3, Mouse Monoclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Brand: Epredia™ MS-1826-S0
Code : LD
Additional Details : Weight : 0.00170kg
Description
Human papilloma virus is non-enveloped double stranded circular DNA virus with icosahedral capsid.Host Species: Mouse
Clone: K1H8
Isotype: IgG1/κ
Species Reactivity: Human Papilloma Virus type 6, 11, 16, 18, 31, 33, 42, 51, 52, 56 and 58.
Epitope: Nonconfirmational internal linear epitope of major capsid protein
Immunogen: Alkaline disrupted HPV type 1
Molecular Weight: 57kDa (major band)
Positive Control: Infected cells and tissue
Cellular Localization: Nuclei of infected cells
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Human Papilloma Virus (HPV) Ab-3 | |
Monoclonal | |
Unconjugated | |
Alkaline disrupted HPV type 1 | |
100 μL | |
Infectious Disease | |
Human, Virus | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
K1H8 | |
Mouse | |
Infected cells and tissue | |
ASR, IVD | |
Primary | |
Concentrated |