missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Human Papilloma Virus (HPV) Ab-3 Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Human Papilloma Virus (HPV) Ab-3, Mouse Monoclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Brand:  Epredia™ MS-1826-S

Code : LD

Additional Details : Weight : 0.00250kg

 View more versions of this product

Product Code. 12610227

  • £505.00 / 1mL
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Human papilloma virus is non-enveloped double stranded circular DNA virus with icosahedral capsid.

Host Species: Mouse

Clone: K1H8

Isotype: IgG1/κ

Species Reactivity: Human Papilloma Virus type 6, 11, 16, 18, 31, 33, 42, 51, 52, 56 and 58.

Epitope: Nonconfirmational internal linear epitope of major capsid protein

Immunogen: Alkaline disrupted HPV type 1

Molecular Weight: 57kDa (major band)

Positive Control: Infected cells and tissue

Cellular Localization: Nuclei of infected cells

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Human Papilloma Virus (HPV) Ab-3
Monoclonal
Unconjugated
Alkaline disrupted HPV type 1
1 mL
Infectious Disease
Human, Virus
IgG2a κ
Immunohistochemistry (Paraffin)
K1H8
Mouse
Infected cells and tissue
ASR, IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers