missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ HER-2/c-erbB-2/neu Rabbit Monoclonal Antibody supernatant
Shop All Epredia ProductsDescription
HER2 gene encodes a transmembrane glycoprotein with tyrosine kinase activity. It is closely related in structure to the epidermal growth factor receptor c-erbB-3/HER3/neu oncoprotein. The protein is detectable in a subset of breast and other carcinomas (urinary bladder, stomach, lung, endometrium and ovary). In the case of breast cancer, over expression determined by immunohistochemistry has been shown to be associated with poor prognosis and an indication to initiate trastuzumab/herceptin therapy.
Host Species: Rabbit
Clone: EP1045Y
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: A synthetic peptide corresponding to the c-terminus of human Her2
Molecular Weight: 185 kDa
Positive Control: Breast carcinomas
Cellular Localization: Cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | HER-2/c-erbB-2/neu |
| Applications | Immunohistochemistry |
| Clone | EP1045Y |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to the c-terminus of human Her2 |
| Quantity | 1 mL |
| Primary or Secondary | Primary |
| Target Species | Human |
| Form | Supernatant |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?